Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145708.2 | NP_663754.2 | 4 | 1176 | ||
GenBank mRNA | BC094912.1 | - | 4 | 1056 | ||
DV647958.1 | - | 1 | 564 |
Gene Symbol: Obox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027802.2 | NP_082078.1 | 4 | 1084 | ||
GenBank mRNA | AK018362.1 | - | 4 | 1081 | ||
AK136061.1 | - | 4 | 1074 |