Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028006.2 | NP_082282.1 | 10 | 1043 | ||
GenBank mRNA | AK010005.1 | - | 10 | 1037 | ||
AK040492.1 | - | 2 | 2601 | |||
AK041416.1 | - | 10 | 1009 | |||
AK041986.1 | - | 449 | ||||
AK087403.1 | - | 9 | 822 | |||
AK148039.1 | - | 10 | 1043 | |||
BC038654.1 | - | 7 | 1008 | |||
BC062179.1 | - | 10 | 1040 | 6333817 |