Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028176.1 | NP_082452.1 | 4 | 595 | ||
GenBank mRNA | AK008793.1 | - | 4 | 601 | ||
BC022649.1 | - | 2 | 322 | |||
BC050114.1 | - | 4 | 595 | 6433052 | ||
BC056179.1 | - | 3 | 431 |