Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028295.1 | NP_082571.1 | 4 | 372 | ||
GenBank mRNA | BC009151.1 | - | 4 | 372 | 3600849 |