Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028312.3 | NP_082588.2 | 5, 6 | 402 | ||
GenBank mRNA | AK012610.1 | - | 5, 6 | 403 | ||
BC026668.1 | - | 5, 6 | 352 | 5006213 | ||
BE627582.1 | - | 1, 2 | 137 | |||
CA450868.1 | - | 3, 4 | 136 |