Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029293.2 | NP_083569.1 | 1 | 52 | ||
GenBank mRNA | AK005756.1 | - | 1 | 52 | ||
AK154580.1 | - | 1 | 73 | |||
BC028657.1 | - | 1 | 25 | 1246740 |