Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029529.3 | NP_083805.1 | 2 | 1542 | ||
GenBank mRNA | AK018094.1 | - | 1 | 324 | ||
AK042761.1 | - | 2 | 1294 | |||
AK044973.1 | - | 2 | 1279 | |||
AK047610.1 | - | 2 | 1543 | |||
BC139194.1 | - | 2 | 1372 | 8862216 |