Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam159b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029984.1 | NP_084260.1 | 3 | 561 | ||
GenBank mRNA | AK020882.1 | - | 3 | 425 | ||
AK042897.1 | - | 3 | 561 |