Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 16 | 2742 | ||
GenBank mRNA | AK050813.1 | - | 14 | 2175 | ||
AK085476.1 | - | 12 | 1770 | |||
AK169344.1 | - | 16 | 2738 | |||
AK170404.1 | - | 16 | 2745 | |||
AK170585.1 | - | 16 | 2665 | |||
AK171148.1 | - | 16 | 2697 | |||
AK171426.1 | - | 16 | 2723 | |||
BC019389.1 | - | 16 | 2545 | 3663506 | ||
X94616.1 | - | 16 | 2590 |