Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 10 | 1424 | ||
NM_001039352.2 | NP_001034441.1 | 10 | 1430 | |||
NM_001039353.2 | NP_001034442.1 | 10 | 1427 | |||
NM_053086.3 | NP_444316.2 | 10 | 1427 | |||
GenBank mRNA | AK014336.1 | - | 10 | 1329 | ||
AK029150.1 | - | 10 | 1275 | |||
AK034817.1 | - | 10 | 1304 | |||
AK129040.1 | - | 10 | 1254 | |||
AK147118.1 | - | 10 | 1300 | |||
AK150332.1 | - | 10 | 1261 | |||
AK151031.1 | - | 10 | 1261 | |||
AK152941.1 | - | 10 | 1261 | |||
AK166753.1 | - | 10 | 1333 | |||
BC003244.1 | - | 9 | 1392 |