Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134438.3 | NP_602320.2 | 2 | 1796 | ||
GenBank mRNA | AB016602.1 | - | 2 | 1539 | ||
AK158275.1 | - | 2 | 1799 | |||
BC098482.1 | - | 2 | 1487 | 30103328 | ||
BC127619.1 | - | 2 | 1420 | 40106370 | ||
BC127620.1 | - | 2 | 1420 | 40106375 | ||
U80895.1 | - | 1 | 849 |
Set Membership: |
G-protein coupled receptor |