Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031363.1 | - | 7 | 982 | ||
AK045891.1 | - | 7 | 965 | |||
AK050529.1 | - | 1 | 1466 | |||
AK053924.1 | - | 7 | 983 | |||
AK078035.1 | - | 5 | 735 | |||
AK078867.1 | - | 7 | 941 | |||
AK083431.1 | - | 5 | 746 | |||
AK181011.1 | - | 1 | 36 | |||
AK181420.1 | - | 1 | 36 | |||
BC012260.1 | - | 7 | 942 | |||
BC128314.1 | - | 6 | 727 | |||
BC131928.1 | - | 7 | 846 | 40130205 | ||
BC131930.1 | - | 7 | 846 | 40130207 |