Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020824.3 | NP_065875.3 | 8, 9 | 1005 | ||
XM_005252542.3 | XP_005252599.1 | 8, 9 | 1073 | |||
XM_011519602.2 | XP_011517904.1 | 8, 9 | 2758 | |||
XM_011519603.2 | XP_011517905.1 | 8, 9 | 1165 | |||
XM_011519604.2 | XP_011517906.1 | 8, 9 | 981 | |||
XM_011519606.2 | XP_011517908.1 | 5, 6 | 2623 | |||
XM_011519607.2 | XP_011517909.1 | 8, 9 | 2758 | |||
XM_017016459.1 | XP_016871948.1 | 8, 9 | 1165 | |||
GenBank mRNA | AB037845.2 | - | 7, 8 | 478 | ||
AF480466.1 | - | 8, 9 | 952 | |||
AK295649.1 | - | 1, 2 | 95 | |||
AK307604.1 | - | 8, 9 | 982 |