Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 3 | 584 | ||
NM_001319652.1 | NP_001306581.1 | 6 | 709 | |||
NM_001319653.1 | NP_001306582.1 | 4 | 621 | |||
NM_020841.4 | NP_065892.1 | 4 | 630 | |||
XM_005268621.4 | XP_005268678.1 | 7 | 728 | |||
XM_006719224.2 | XP_006719287.1 | 3 | 589 | |||
XM_011537855.2 | XP_011536157.1 | 4 | 247 | |||
XM_011537856.2 | XP_011536158.1 | 5 | 665 | |||
XM_011537857.2 | XP_011536159.1 | 7 | 719 | |||
XM_017018768.1 | XP_016874257.1 | 4 | 262 | |||
XM_017018769.1 | XP_016874258.1 | 5 | 633 | |||
GenBank mRNA | AB040884.3 | - | 4 | 301 | ||
AF323730.1 | - | 3 | 584 | |||
AF392452.1 | - | 4 | 631 | |||
AK055835.1 | - | 3 | 182 | |||
AK289997.1 | - | 4 | 350 | |||
BC082235.1 | - | 4 | 569 | |||
BC093834.1 | - | 3 | 229 | 7939679 | ||
BC101529.1 | - | 3 | 229 | 8069035 | ||
BC111728.1 | - | 4 | 524 | 40029118 | ||
BX647211.1 | - | 3 | 183 | |||
CR749542.1 | - | 6 | 435 |