Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286725.1 | NP_001273654.1 | 1 | 1273 | ||
NM_020866.2 | NP_065917.1 | 1 | 1273 | |||
GenBank mRNA | AB040923.1 | - | 1 | 1239 | ||
AF252283.1 | - | 1 | 1107 | |||
AK054609.1 | - | 1 | 485 | |||
AK291435.1 | - | 1 | 1273 | |||
AK295939.1 | - | 1 | 464 | |||
BC022460.1 | - | 1 | 517 | 4819077 |