Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146684.1 | NP_666895.1 | 1 | 304 | ||
GenBank mRNA | BC125000.1 | - | 1 | 305 |