Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024512.2 | NP_001019683.1 | 7 | 998 | ||
NM_001177633.1 | NP_001171104.1 | 7 | 998 | |||
NM_172379.3 | NP_759011.2 | 5 | 719 | |||
GenBank mRNA | AK031973.1 | - | 6 | 839 | ||
AK046169.1 | - | 1 | 82 | |||
AK077495.1 | - | 7 | 901 | |||
AK129252.1 | - | 5 | 623 | |||
BC008267.1 | - | 1 | 62 | |||
BC025183.1 | - | 7 | 894 | 5011464 | ||
BC057907.1 | - | 5 | 621 | 5064176 | ||
BC069846.1 | - | 7 | 896 | 4196478 | ||
BC080849.1 | - | 7 | 938 | 5717789 |