Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173742.3 | NP_776103.1 | 3 | 316 | ||
GenBank mRNA | AK003548.1 | - | 3 | 292 | ||
AK009592.1 | - | 3 | 311 | |||
AK018709.1 | - | 3 | 265 | |||
AK171975.1 | - | 3 | 265 | |||
BC021602.1 | - | 3 | 244 | 5102170 | ||
BC021603.1 | - | 3 | 244 | 5102465 | ||
BC024417.1 | - | 3 | 274 | 5065541 | ||
CJ181610.1 | - | 3 | 316 |