Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174865.1 | NP_777354.1 | 5 | 733 | ||
GenBank mRNA | BC106965.1 | - | 5 | 733 | 40039744 | |
BC106966.2 | - | 5 | 730 | |||
BC107012.1 | - | 5 | 733 | 40047687 |