Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbq1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175000.2 | NP_778165.1 | 3 | 420 | ||
GenBank mRNA | AK039049.1 | - | 3 | 421 | ||
BC139349.1 | - | 3 | 394 | 8862371 | ||
BC139351.1 | - | 3 | 394 | 8862373 |