Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039959.2 | NP_001035048.1 | 4 | 475 | ||
NM_001286518.1 | NP_001273447.1 | 4 | 475 | |||
NM_009643.2 | NP_033773.1 | 4 | 475 | |||
GenBank mRNA | AK009866.1 | - | 4 | 498 | ||
AK045184.1 | - | 4 | 476 | |||
AK134304.1 | - | 4 | 1487 | |||
BC028439.1 | - | 3 | 340 | |||
CB321491.1 | - | 4 | 458 |