Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175167.3 | NP_780376.2 | 2 | 160 | ||
GenBank mRNA | AK029377.1 | - | 2 | 186 | ||
BC047134.1 | - | 2 | 160 | 6308426 | ||
BC080814.1 | - | 2 | 169 | 6391119 |