Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021449.3 | NP_067424.2 | 11 | 1528 | ||
NM_175357.3 | NP_780566.1 | 11 | 1531 | |||
GenBank mRNA | AF229032.1 | - | 11 | 1483 | ||
AK032836.1 | - | 11 | 1505 | |||
AK050557.1 | - | 11 | 1514 | |||
AK075959.1 | - | 11 | 1504 | |||
AK076144.1 | - | 11 | 1511 | |||
AK077707.1 | - | 11 | 1511 | |||
AK160219.1 | - | 11 | 1507 | |||
BC033458.1 | - | 11 | 1489 | |||
BC046967.1 | - | 10 | 1446 | |||
BC069905.1 | - | 11 | 1488 | |||
BC086488.1 | - | 11 | 1506 | 30443656 |