Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28818
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
4
520
NM_001007471.2
NP_001007472.2
4
868
NM_020952.4
NP_066003.3
4
520
NM_024971.5
NP_079247.5
4
520
NM_206944.3
NP_996827.3
4
520
NM_206945.3
NP_996828.3
4
520
NM_206946.3
NP_996829.3
4
520
NM_206947.3
NP_996830.3
4
520
NM_206948.2
NP_996831.1
4
520
XM_011519035.2
XP_011517337.1
4
745
XM_011519036.2
XP_011517338.1
4
3670
XM_011519037.2
XP_011517339.1
4
3656
XM_011519038.2
XP_011517340.1
4
3653
XM_011519039.2
XP_011517341.1
4
3654
XM_011519040.2
XP_011517342.1
4
751
XM_011519041.2
XP_011517343.1
4
3653
XM_011519042.2
XP_011517344.1
4
751
XM_011519043.2
XP_011517345.1
4
784
XM_011519044.2
XP_011517346.1
4
3654
XM_011519045.2
XP_011517347.1
4
745
XM_011519046.2
XP_011517348.1
4
2448
XM_011519047.2
XP_011517349.1
4
2418
XM_017015141.1
XP_016870630.1
4
745
XM_017015142.1
XP_016870631.1
4
745
XM_017015143.1
XP_016870632.1
4
745
XM_017015144.1
XP_016870633.1
4
511
XM_017015145.1
XP_016870634.1
4
1189
XM_017015146.1
XP_016870635.1
4
587
XM_017015147.1
XP_016870636.1
4
2622
XM_017015148.1
XP_016870637.1
4
745
XM_017015149.1
XP_016870638.1
4
745
XM_017015150.1
XP_016870639.1
4
745
XM_017015151.1
XP_016870640.1
4
763
XM_017015152.1
XP_016870641.1
4
2423
XM_017015153.1
XP_016870642.1
4
755
XM_017015154.1
XP_016870643.1
4
891
XM_017015155.1
XP_016870644.1
4
2420
XM_017015157.1
XP_016870646.1
4
2418
XM_017015158.1
XP_016870647.1
4
2416
XM_017015159.1
XP_016870648.1
4
511
XM_017015160.1
XP_016870649.1
4
601
XM_017015161.1
XP_016870650.1
4
745
GenBank mRNA
AB099661.1
-
1
165
AB099662.1
-
1
165
AB099663.1
-
1
165
AB099664.1
-
1
165
AB099665.1
-
1
165
AF536748.1
-
1
165
AF536749.1
-
1
165
AF536750.1
-
1
165
AF536751.1
-
1
165
AF536752.1
-
1
165
AF536753.1
-
1
165
AJ505025.1
-
4
868
AJ505026.1
-
4
868
AK225732.1
-
4
511
AL136545.1
-
4
701
BC094699.1
-
4
514
CD104154.1
-
4
516
KF987075.1
-
4
576
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0