Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175450.4 | NP_780659.2 | 7 | 886 | ||
GenBank mRNA | AK050106.1 | - | 7 | 884 | ||
AK082695.1 | - | 7 | 889 | |||
AK133716.1 | - | 7 | 887 | |||
AK160713.1 | - | 7 | 884 | |||
AK166113.1 | - | 884 | ||||
AK168691.1 | - | 7 | 884 | |||
BC032968.1 | - | 6 | 578 | |||
BC095984.1 | - | 7 | 886 | 4978914 |