Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176996.4 | NP_795970.3 | 12 | 2535 | ||
GenBank mRNA | AK081050.1 | - | 6 | 723 | ||
BC030377.1 | - | 5 | 572 | |||
BC096028.1 | - | 12 | 2054 | 6850065 | ||
FJ883552.1 | - | 12 | 2044 |
Set Membership: |
G-protein coupled receptor |