Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177361.2 | NP_796335.1 | 1 | 678 | ||
GenBank mRNA | AY190046.1 | - | 1 | 678 | ||
BC104352.1 | - | 1 | 657 | 40049569 | ||
BC104353.1 | - | 1 | 657 | 40049571 |