Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177362.3 | NP_796336.1 | 2 | 205 | ||
GenBank mRNA | AK090240.1 | - | 2 | 206 | ||
BC139048.1 | - | 2 | 188 | 8862070 | ||
BC139049.1 | - | 2 | 188 | 8862071 | ||
BC145524.1 | - | 2 | 201 | 9054087 |
Gene Symbol: Mlph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC006900.1 | - | 2471 |