Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252519.1 | NP_001239448.1 | 6 | 1980 | ||
NM_177889.5 | NP_808557.2 | 7 | 2070 | |||
GenBank mRNA | AK129467.1 | - | 2 | 3481 | ||
AK137630.1 | - | 1694 | ||||
BC025629.1 | - | 1 | 1226 | |||
BC060530.1 | - | 6 | 1997 | 5684819 | ||
BX525285.1 | - | 1 | 373 |