Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252519.1 | NP_001239448.1 | 6 | 769 | ||
NM_177889.5 | NP_808557.2 | 7 | 859 | |||
GenBank mRNA | AK129467.1 | - | 2 | 2270 | ||
AK137630.1 | - | 483 | ||||
BC060530.1 | - | 6 | 786 | 5684819 | ||
CD806198.1 | - | 5 | 720 |