Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 4 | 468 | ||
NM_001190323.2 | NP_001177252.1 | 4 | 344 | |||
NM_001190324.2 | NP_001177253.1 | 4 | 299 | |||
NM_001285804.1 | NP_001272733.1 | 4 | 499 | |||
NM_181391.4 | NP_852056.1 | 4 | 513 | |||
GenBank mRNA | AA980189.1 | - | 4 | 296 | ||
AK003308.1 | - | 4 | 345 | |||
AK007844.1 | - | 4 | 488 | |||
AK013200.1 | - | 4 | 486 | |||
AK145105.1 | - | 4 | 488 | |||
AK168647.1 | - | 4 | 443 | |||
AK169018.1 | - | 4 | 488 | |||
AV042014.2 | - | 2 | 108 | |||
BC030366.1 | - | 4 | 471 | 5370748 | ||
BX527959.1 | - | 4 | 260 | |||
CB057699.1 | - | 4 | 425 |