Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293795.1 | NP_001280724.1 | 10 | 1095 | ||
NM_181540.4 | NP_853518.1 | 10 | 1089 | |||
GenBank mRNA | AK077126.1 | - | 10 | 1082 | ||
BC024498.1 | - | 10 | 1083 | 4982620 |