Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181596.4 | NP_853627.2 | 4 | 511 | ||
GenBank mRNA | AB086819.1 | - | 3 | 432 | ||
AJ514933.1 | - | 3 | 435 | |||
AJ536019.1 | - | 4 | 474 | |||
AK158401.1 | - | 4 | 475 | |||
BC117106.1 | - | 3 | 424 | 40126425 |