Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183194.3 | NP_899017.2 | 7, 8 | 911 | ||
GenBank mRNA | AK037079.1 | - | 7, 8 | 912 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC099685.1 | - | 955 | Chr. - - [-] on Build |