Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx6-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183248.3 | NP_899071.2 | 3 | 1021 | ||
GenBank mRNA | AK008173.1 | - | 3 | 1022 | ||
L08074.1 | - | 3 | 959 |