Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183257.3 | NP_899080.1 | 3 | 367 | ||
GenBank mRNA | AK007975.1 | - | 3 | 357 | ||
AK008993.1 | - | 3 | 368 | |||
AY232841.1 | - | 3 | 356 | |||
BC086895.1 | - | 3 | 364 | 6765293 | ||
D17962.1 | - | 1 | 172 |