Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197991.2 | NP_932108.2 | 7 | 1493 | ||
GenBank mRNA | AK009801.1 | - | 7 | 1410 | ||
AK150499.1 | - | 7 | 1398 | |||
AK160274.1 | - | 7 | 1412 | |||
AK169010.1 | - | 7 | 1444 | |||
AK171657.1 | - | 7 | 1429 | |||
AY761096.1 | - | 8 | 1473 | |||
AY761098.1 | - | 7 | 1386 | |||
BC006065.1 | - | 5 | 1146 | |||
BC020179.1 | - | 7 | 1410 | 4913500 | ||
BY761533.1 | - | 1 | 273 |