Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021002.2 | NP_066282.1 | 1 | 254 | ||
GenBank mRNA | BC069471.1 | - | 1 | 254 | 7262180 | |
BC096697.1 | - | 1 | 254 | 40014596 | ||
BC096710.1 | - | 1 | 254 | 40014597 | ||
BC096730.1 | - | 1 | 254 | 40014598 | ||
BC098357.1 | - | 1 | 254 | 40014594 |