Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172099.1 | NP_001165570.1 | 4, 5 | 1068 | ||
NM_198013.3 | NP_932130.2 | 5, 6 | 1266 | |||
GenBank mRNA | AK049250.1 | - | 5, 6 | 1230 | ||
AK162604.1 | - | 4, 5 | 648 | |||
BC024476.1 | - | 5, 6 | 1115 | 4976954 | ||
BC024529.1 | - | 1, 2 | 67 | |||
BC025434.1 | - | 5, 6 | 1120 | |||
BC029708.1 | - | 5, 6 | 1199 | 5041450 | ||
BC131989.1 | - | 4, 5 | 1039 | 40130266 | ||
BC131991.1 | - | 4, 5 | 1039 | 40130268 | ||
BC145206.1 | - | 4, 5 | 1039 | 9053757 | ||
CF950088.1 | - | 1, 2 | 128 | |||
CK332869.1 | - | 4, 5 | 512 |