Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF067061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199060.1 | NP_951015.1 | 582 | |||
GenBank mRNA | AF067061.1 | - | 581 | |||
AF067062.1 | - | 582 | ||||
BC115951.1 | - | 1 | 1028 | 40049464 |
Gene Symbol: Gm20767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039648.3 | NP_001034737.2 | 3 | 1129 | ||
GenBank mRNA | AF067058.1 | - | 2 | 577 | ||
AK139926.1 | - | 3 | 1130 | |||
BC147576.1 | - | 2 | 609 | 9056566 | ||
BC147578.1 | - | 2 | 609 | 9056568 |