Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021010.2 | NP_066290.1 | 1 | 119 | ||
GenBank mRNA | BC069690.1 | - | 1 | 115 | 7262486 | |
BC107079.2 | - | 1 | 110 | 40014027 | ||
BP419449.1 | - | 1 | 64 |