Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077064.1 | - | 4 | 178 | ||
FJ541086.1 | - | 227 | ||||
FJ541087.1 | - | 159 | ||||
FJ541088.1 | - | 159 | ||||
FJ541094.1 | - | 92 | ||||
FJ541095.1 | - | 92 |
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ541076.1 | - | 291 |
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 5 | 298 | ||
GenBank mRNA | BC049626.1 | - | 5 | 298 | 6705850 |