Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201641.2 | NP_964003.2 | 1 | 596 | ||
GenBank mRNA | AY227201.1 | - | 1 | 555 | ||
BC089569.1 | - | 1 | 596 | 30120494 |