Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 4 | 340 | ||
XM_011543885.2 | XP_011542187.1 | 4 | 293 | |||
GenBank mRNA | AK310348.1 | - | 4 | 282 | ||
AK310513.1 | - | 4 | 293 | |||
BC005904.1 | - | 4 | 294 | 4064072 | ||
BC103862.1 | - | 4 | 339 | 40003336 | ||
DB444418.1 | - | 4 | 401 | |||
S82471.1 | - | 3 | 200 | |||
U90840.1 | - | 4 | 253 |