Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201645.2 | NP_964007.2 | 1 | 590 | ||
GenBank mRNA | AY227194.1 | - | 1 | 590 | ||
BC093516.1 | - | 1 | 552 | 5135493 | ||
JX026002.1 | - | 1 | 547 | |||
L02333.1 | - | 1 | 562 | |||
S64760.1 | - | 1 | 562 |