Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206975.1 | NP_996858.1 | 1 | 556 | ||
GenBank mRNA | BC120722.1 | - | 1 | 606 | 40129645 | |
BC125321.1 | - | 1 | 606 | 40129836 |