Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 6 | 1398 | ||
XM_011527126.1 | XP_011525428.1 | 5 | 1119 | |||
XM_011527127.1 | XP_011525429.1 | 4 | 864 | |||
GenBank mRNA | BC005924.1 | - | 6 | 1363 | 4043628 | |
M23575.1 | - | 6 | 1497 | |||
M27907.1 | - | 3 | 482 | |||
M30628.1 | - | 8 | 1426 | |||
M34420.1 | - | 6 | 1347 | |||
M37399.1 | - | 6 | 1352 | |||
M93061.1 | - | 6 | 1324 | |||
S59493.1 | - | 6 | 1324 | |||
S59494.1 | - | 8 | 1432 |