Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207105.3 | NP_996988.2 | 6 | 1017 | ||
GenBank mRNA | AK077436.1 | - | 6 | 1005 | ||
AK155573.1 | - | 6 | 989 | |||
AK156899.1 | - | 6 | 989 | |||
AK170370.1 | - | 6 | 999 | |||
AK171005.1 | - | 6 | 989 | |||
AK171211.1 | - | 6 | 993 | |||
AK171281.1 | - | 6 | 989 | |||
AK171290.1 | - | 6 | 989 | |||
AK171298.1 | - | 6 | 989 | |||
AK172409.1 | - | 6 | 990 | |||
BC008168.1 | - | 6 | 1009 | 3257494 | ||
BC010322.1 | - | 6 | 983 | 2651058 | ||
K01143.1 | - | 3 | 292 |