Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021033.6 | NP_066361.1 | 1 | 384 | ||
GenBank mRNA | AF205602.1 | - | 1 | 151 | ||
AF493914.1 | - | 1 | 134 | |||
AK315139.1 | - | 2 | 180 | |||
BC041333.2 | - | 1 | 385 | 5278625 | ||
BC070031.1 | - | 1 | 142 | 4815530 | ||
DB480985.1 | - | 1 | 439 | |||
X12534.1 | - | 1 | 137 |