Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021049.4 | NP_066387.1 | 3 | 422 | ||
GenBank mRNA | AY310324.1 | - | 1 | 250 | ||
BC109187.1 | - | 3 | 422 | 40000127 |
Gene Symbol: MAGEA10-MAGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204811.2 | NP_001191740.1 | 4 | 631 |